missing translation for 'onlineSavingsMsg'
Learn More

E-64, MedChemExpress

Product Code. 16476418
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16476418 10 mg 10mg
16473466 25 mg 25mg
16459007 5 mg 5mg
16432527 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16476418 Supplier Medchem Express Supplier No. HY15282
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress E-64 (Proteinase inhibitor E 64) is a potent irreversible inhibitor against general cysteine proteases with IC50 of 9 nM for papain.

E-64 (Proteinase inhibitor E 64) is a potent irreversible inhibitor against general cysteine proteases with IC50 of 9 nM for papain.

Specifications

Chemical Name or Material E-64
Physical Form Solid
CAS 66701-25-5
Color White
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C15H27N5O5
For Use With (Application) COVID-19-immunoregulation
Synonym Proteinase inhibitor E 64
Solubility Information DMSO : 100 mg/mL (279.79 mM; Need ultrasonic) ∣H2O : 5 mg/mL (13.99 mM; Need ultrasonic)
SMILES O=C([C@H]1O[C@@H]1C(N[C@H](C(NCCCCNC(N)=N)=O)CC(C)C)=O)O
Molecular Weight (g/mol) 357.41
Formula Weight 357.41
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.