missing translation for 'onlineSavingsMsg'
Learn More

Melford E-64

Product Code. 30018259
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30018259 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30018259 Supplier Melford Supplier No. E570500.005
 Restricted Item
Estimated Shipment: 26-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Irreversible cysteine protease inhibitor of calpain, cathepsin (B,H,L,S), papain and other cysteine proteases.

Irreversible cysteine protease inhibitor of calpain, cathepsin (B,H,L,S), papain and other cysteine proteases. Working concentration: 1-10uM.

Specifications

CAS 66701-25-5
Molecular Formula C15H27N5O5
Quantity 5 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.