missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ E 64

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
E 64 is a potent and irreversible cysteine protease inhibitor (IC50 values are 1.4, 2.5 and 4.1 nM for cathepsin K, L and S respectively. Also inhibits papain, calpain, cathepsin B and cathepsin H. Has no effect on serine proteases. Reduces HSC-3 tongue cancer cell invasion and induces oxidative stress and apoptosis in filarial parasites. Also inhibits autophagy-associated lysosomal degradation in vitro.
Specifications
Specifications
| Quantity | 10 mg |
| Product Type | E 64 |
| Purity | >98% |
| Molecular Weight (g/mol) | 357.41 |
| Inhibitors | E 64 |
For Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction