missing translation for 'onlineSavingsMsg'
Learn More

(E)-(5,6-Dihydro-1,4,2-dioxazin-3-yl)[2-[(tetrahydro-2H-pyran-2-yl)oxy]phenyl]-methanone O-Methyloxime-d4, TRC

Product Code. 30385450
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30385450 5 mg 5mg
30376118 10 mg 10mg
30381853 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30385450 Supplier Toronto Research Chemicals Supplier No. TRCD4866865MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (E)-(5,6-Dihydro-1,4,2-dioxazin-3-yl)[2-[(tetrahydro-2H-pyran-2-yl)oxy]phenyl]-methanone O-Methyloxime-d4
Molecular Formula C16 D4 H16 N2 O5
Quantity 5 mg
InChI Formula InChI=1S/C16H20N2O5/c1-19-17-15(16-18-22-11-10-21-16)12-6-2-3-7-13(12)23-14-8-4-5-9-20-14/h2-3,6-7,14H,4-5,8-11H2,1H3/b17-15+/i10D2,11D2
SMILES [2H]C1([2H])ON=C(OC1([2H])[2H])\C(=N\OC)\c2ccccc2OC3CCCCO3
IUPAC Name (E)-N-methoxy-1-[2-(oxan-2-yloxy)phenyl]-1-(5,5,6,6-tetradeuterio-1,4,2-dioxazin-3-yl)methanimine
Molecular Weight (g/mol) 324.365
Formula Weight 324.162
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.