missing translation for 'onlineSavingsMsg'
Learn More

E-4031 dihydrochloride, TRC

Product Code. 30384377
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30384377 5 mg 5mg
30387914 10 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30384377 Supplier Toronto Research Chemicals Supplier No. TRCD4539085MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 113559-13-0
Molecular Formula C21H27N3O3S • 2HCl
Molecular Weight (g/mol) 401.5223646
Synonym E-4031
SMILES O=C(C1=CC=C(NS(=O)(C)=O)C=C1)C(CC2)CCN2CCC3=NC(C)=CC=C3.Cl

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C21H27N3O3S.2ClH/c1-16-4-3-5-19(22-16)12-15-24-13-10-18(11-14-24)21(25)17-6-8-20(9-7-17)23-28(2,26)27;;/h3-9,18,23H,10-15H2,1-2H3;2*1H
Chemical Name or Material E-4031 Dihydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.