missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ E-4031 dihydrochloride

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
Selective blocker of KV11.1 (hERG) channels; inhibits the rapid delayed-rectifier K+ current (IKr). Reversibly prolongs action potential duration in guinea pig papillary muscle and isolated ventricular myocytes, without affecting Na+ or Ca2+ inward currents. Class III antiarrhythmic agent.
Specifications
Specifications
| Quantity | 50 mg |
| Formula Weight | 474.44 |
| Purity | >98% |
| Molecular Weight (g/mol) | 474.44 |
| Chemical Name or Material | E-4031 dihydrochloride |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction