missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ E-4031 dihydrochloride
GREENER_CHOICE

Product Code. 18044692
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18044692

Brand: Tocris Bioscience™ 1808/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

KV11.1 (hERG) channel blocker; inhibits rapid delayed rectifier K+ current (IKr)

Selective blocker of KV11.1 (hERG) channels; inhibits the rapid delayed-rectifier K+ current (IKr). Reversibly prolongs action potential duration in guinea pig papillary muscle and isolated ventricular myocytes, without affecting Na+ or Ca2+ inward currents. Class III antiarrhythmic agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 113559-13-0
Molecular Formula C21H29Cl2N3O3S

Specifications

Quantity 10 mg
Formula Weight 474.44
Purity >98%
Molecular Weight (g/mol) 474.44
Chemical Name or Material E-4031 dihydrochloride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.