missing translation for 'onlineSavingsMsg'
Learn More

(E)-4-Hydroxycrotonoic Acid Ethyl Ester (~85%), TRC

Product Code. 30388916
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30388916 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30388916 Supplier Toronto Research Chemicals Supplier No. TRCH918465250MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (E)-4-Hydroxycrotonoic Acid Ethyl Ester
CAS 10080-68-9
Molecular Formula C6H10O3
Quantity 250 mg
Synonym Ethyl trans-4-Hydroxy-2-butenoate, (2E)-4-Hydroxy-2-butenoic Acid Ethyl Ester
InChI Formula InChI=1S/C6H10O3/c1-2-9-6(8)4-3-5-7/h3-4,7H,2,5H2,1H3/b4-3+
SMILES CCOC(=O)\C=C\CO
IUPAC Name ethyl (E)-4-hydroxybut-2-enoate
Molecular Weight (g/mol) 130.14
Formula Weight 130.063
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.