missing translation for 'onlineSavingsMsg'
Learn More

(E)-4-Hydroxy-N-desmethyl Tamoxifen, TRC

Product Code. 30397240
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30397240 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30397240 Supplier Toronto Research Chemicals Supplier No. TRCH93849510MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (E)-4-Hydroxy-N-desmethyl Tamoxifen
CAS 114828-90-9
Molecular Formula C25H27NO2
Quantity 10 mg
InChI Formula InChI=1S/C25H27NO2/c1-3-24(19-7-5-4-6-8-19)25(20-9-13-22(27)14-10-20)21-11-15-23(16-12-21)28-18-17-26-2/h4-16,26-27H,3,17-18H2,1-2H3/b25-24+
SMILES CC\C(=C(\c1ccc(O)cc1)/c2ccc(OCCNC)cc2)\c3ccccc3
IUPAC Name 4-[(E)-1-[4-[2-(methylamino)ethoxy]phenyl]-2-phenylbut-1-enyl]phenol
Molecular Weight (g/mol) 373.49
Formula Weight 373.2042
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.