missing translation for 'onlineSavingsMsg'
Learn More

E-4-(Dimethylamino)cinnamaldehyde, TRC

Product Code. 30385935
Change view
Click to view available options
Quantity:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30385935 10 mg
30387630 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30385935 Supplier Toronto Research Chemicals Supplier No. TRCD49361410MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20432-35-3
Molecular Formula C11H13NO
Molecular Weight (g/mol) 175.23
Synonym 4-(Dimethylamino)-trans-cinnamaldehyde, (E)-3-[4-(Dimethylamino)phenyl]-2-propenal, (E)-3-(4-Dimethylaminophenyl)-prop-2-enal, trans-p-(Dimethylamino)cinnamaldehyde
SMILES CN(C)c1ccc(\C=C\C=O)cc1

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C11H13NO/c1-12(2)11-7-5-10(6-8-11)4-3-9-13/h3-9H,1-2H3/b4-3+
Formula Weight 175.0997
Chemical Name or Material E-4-(Dimethylamino)cinnamaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.