missing translation for 'onlineSavingsMsg'
Learn More

(E)-4,5-Dihydroxypent-2-enoic Acid Sodium Salt, TRC

Product Code. 30374485
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30374485 50 mg 50mg
30376618 10 mg 10mg
30382465 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30374485 Supplier Toronto Research Chemicals Supplier No. TRCD80139050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (E)-4,5-Dihydroxypent-2-enoic Acid Sodium Salt
Analyte or Component Names (E)-4,5-Dihydroxypent-2-enoic Acid Sodium Salt
Shipping Condition Room Temperature
Molecular Formula C5H7O4Na
Synonym Sodium (E)-4,5-Dihydroxypent-2-enoate
InChI Formula InChI=1S/C5H8O4.Na/c6-3-4(7)1-2-5(8)9;/h1-2,4,6-7H,3H2,(H,8,9);/q;+1/p-1/b2-1+;
Solution Type Neat
SMILES O=C(O[Na])/C=C/C(O)CO
Industry Type Pharmaceutical
Molecular Weight (g/mol) 154.1
Quantity 50 mg
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.