missing translation for 'onlineSavingsMsg'
Learn More

(E)-4-(3,5-Dimethoxystyryl)benzene-1,2-diol, TRC

Product Code. 30382076
Change view
Click to view available options
Quantity:
1 mg
2.5 mg
5 mg
Unit Size:
2.50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30382076 2.5 mg 2.50mg
30377742 1 mg -
30386927 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30382076 Supplier Toronto Research Chemicals Supplier No. TRCD4791982.5MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 475231-21-1
Molecular Formula C16H16O4
Molecular Weight (g/mol) 272.3
Synonym 4-[(1E)-2-(3,5-Dimethoxyphenyl)ethenyl]-1,2-benzenediol, 3'-Hydroxypterostilbene
SMILES COc1cc(OC)cc(\C=C\c2ccc(O)c(O)c2)c1

Specifications

Quantity 2.5 mg
InChI Formula InChI=1S/C16H16O4/c1-19-13-7-12(8-14(10-13)20-2)4-3-11-5-6-15(17)16(18)9-11/h3-10,17-18H,1-2H3/b4-3+
Formula Weight 272.1049
Chemical Name or Material (E)-4-(3,5-Dimethoxystyryl)benzene-1,2-diol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.