missing translation for 'onlineSavingsMsg'
Learn More

(E)-3-(4-((2-Hydroxy-3-(methoxycarbonyl)phenyl)diazenyl)benzamido)propanoic Acid, TRC

Product Code. 30397460
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30397460 10 mg 10mg
30407265 5 mg 5mg
30391319 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30397460 Supplier Toronto Research Chemicals Supplier No. TRCH94614010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (E)-3-(4-((2-Hydroxy-3-(methoxycarbonyl)phenyl)diazenyl)benzamido)propanoic Acid
Analyte or Component Names (E)-3-(4-((2-Hydroxy-3-(methoxycarbonyl)phenyl)diazenyl)benzamido)propanoic Acid
Shipping Condition Room Temperature
Molecular Formula C18 H17 N3 O6
InChI Formula InChI=1S/C18H17N3O6/c1-27-18(26)13-3-2-4-14(16(13)24)21-20-12-7-5-11(6-8-12)17(25)19-10-9-15(22)23/h2-8,24H,9-10H2,1H3,(H,19,25)(H,22,23)/b21-20+
Solution Type Neat
SMILES COC(=O)c1cccc(\N=N\c2ccc(cc2)C(=O)NCCC(=O)O)c1O
Industry Type Pharmaceutical
Molecular Weight (g/mol) 371.344
Quantity 10 mg
Formula Weight 371.112
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.