missing translation for 'onlineSavingsMsg'
Learn More

(E)-3-(3-(Dimethylamino)acryloyl)pyridine 1-Oxide, TRC

Product Code. 30382069
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30382069 10 mg 10mg
30375565 25 mg 25mg
30379786 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30382069 Supplier Toronto Research Chemicals Supplier No. TRCD46210510MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (E)-3-(3-(Dimethylamino)acryloyl)pyridine 1-Oxide
Analyte or Component Names (E)-3-(3-(Dimethylamino)acryloyl)pyridine 1-Oxide
Shipping Condition Room Temperature
Molecular Formula C10 H12 N2 O2
InChI Formula InChI=1S/C10H12N2O2/c1-11(2)7-5-10(13)9-4-3-6-12(14)8-9/h3-8H,1-2H3/b7-5+
Solution Type Neat
SMILES CN(C)\C=C\C(=O)c1ccc[n+]([O-])c1
IUPAC Name (E)-3-(dimethylamino)-1-(1-oxidopyridin-1-ium-3-yl)prop-2-en-1-one
Industry Type Pharmaceutical
Molecular Weight (g/mol) 192.214
Quantity 10 mg
Formula Weight 192.09
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.