missing translation for 'onlineSavingsMsg'
Learn More

(E)-3,3-Diethoxy-1-propenylboronic acid pinacol ester, 97%, Thermo Scientific™

Product Code. 10175847 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10175847 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10175847 Supplier Thermo Scientific Supplier No. 430280010

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 153737-25-8
CAS Min % 96.0
CAS Max % 100.0
Color Colorless
Boiling Point 135.0°C to 145.0°C (0.1 Torr)
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Molecular Formula C13H25BO4
Refractive Index 1.441
MDL Number MFCD03788737
Quantity 1 g
Synonym 3,3-diethoxy-1-propenylboronic acid pinacol ester, e-2-3,3-diethoxyprop-1-en-1-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 3,3-diethoxy-trans-1-propenylboronic acid pinacol ester, 1,3,2-dioxaborolane, 2-3,3-diethoxy-1-propen-1-yl-4,4,5,5-tetramethyl, 2-e-3,3-diethoxy-1-propenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-1e-3,3-diethoxyprop-1-en-1-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, e-3,3-diethoxy-1-propenylboronic acid pinacol ester, 3,3-diethoxy-1-propenylboronic acid picanol ester, e + z, 3-propene aldehyde-boronic acid pinacol ester diethyl acetal, e-2-3,3-diethoxyprop-1-enyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
InChI Key ZXGPKUGXWIEUJA-MDZDMXLPSA-N
SMILES B1(OC(C(O1)(C)C)(C)C)C=CC(OCC)OCC
IUPAC Name 2-[(E)-3,3-diethoxyprop-1-enyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Molecular Weight (g/mol) 256.15
PubChem CID 11065022
Formula Weight 256.15
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material (E)-3, 3-Diethoxy-1-propenylboronic acid pinacol ester
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.