missing translation for 'onlineSavingsMsg'
Learn More

(E)-3-[3-(4-Fluorophenyl)-1-(1-methylethyl)-1H-indol-2-yl]prop-2-enal, Mikromol™

Product Code. 30051849
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Product Code. Quantity unitSize
30051849 25 mg 25mg
1 options
This item is not returnable. View return policy

Product Code. 30051849

Brand: Mikromol MM0676.140025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

(E)-3-[3-(4-Fluorophenyl)-1-(1-methylethyl)-1H-indol-2-yl]prop-2-enal
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (E)-3-[3-(4-Fluorophenyl)-1-(1-methylethyl)-1H-indol-2-yl]prop-2-enal
CAS 93957-50-7
Analyte or Component Names (E)-3-[3-(4-Fluorophenyl)-1-(1-methylethyl)-1H-indol-2-yl]prop-2-enal
Shipping Condition Room Temperature
Molecular Formula C20 H18 F N O
Certifications/Compliance ISO 9001
Synonym 2-Propenal, 3-[3-(4-fluorophenyl)-1-(1-methylethyl)-1H-indol-2-yl]-, (2E)-, 2-Propenal, 3-[3-(4-fluorophenyl)-1-(1-methylethyl)-1H-indol-2-yl]-, (E)-, (2E)-3-[3-(4-Fluorophenyl)-1-(1-methylethyl)-1H-indol-2-yl]-2-propenal
InChI Formula InChI=1S/C20H18FNO/c1-14(2)22-18-7-4-3-6-17(18)20(19(22)8-5-13-23)15-9-11-16(21)12-10-15/h3-14H,1-2H3/b8-5+
SMILES CC(C)n1c(\C=C\C=O)c(c2ccc(F)cc2)c3ccccc13
IUPAC Name (E)-3-[3-(4-fluorophenyl)-1-isopropyl-indol-2-yl]prop-2-enal
Molecular Weight (g/mol) 307.36
Quantity 25 mg
Formula Weight 307.1372 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.