missing translation for 'onlineSavingsMsg'
Learn More

(E)-2-Thiopheneacrylic Acid, TRC

Product Code. 30457129
Change view
Click to view available options
Quantity:
50 g
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30457129 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30457129 Supplier Toronto Research Chemicals Supplier No. TRCT34481550G
 Restricted Item
Estimated Shipment: 11-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (E)-2-Thiopheneacrylic Acid
CAS 15690-25-2
Molecular Formula C7H6O2S
Quantity 50 g
Synonym (E)-3-(Thiophen-2-yl)-2-propenoic Acid, (E)-3-(Thiophen-2-yl)acrylic Acid, trans-3-Thiopheneacrylic Acid, (E)-2-Thiopheneacrylic Acid, (E)-3-(2-Thienyl)-2-propenoic Acid
InChI Formula InChI=1S/C7H6O2S/c8-7(9)4-3-6-2-1-5-10-6/h1-5H,(H,8,9)/b4-3-
SMILES OC(=O)\C=C/c1cccs1
IUPAC Name (Z)-3-thiophen-2-ylprop-2-enoic acid
Molecular Weight (g/mol) 154.19
Formula Weight 154.0089
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.