missing translation for 'onlineSavingsMsg'
Learn More

(E)-2-Hydroxy-5-((4-(N-(4-(N-(pyridin-2-yl)sulfamoyl)phenyl)sulfamoyl)phenyl)diazenyl)benzoic Acid, TRC

Product Code. 30394583
Change view
Click to view available options
Quantity:
25 mg
100 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30394583 25 mg
30403236 100 mg
30405369 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30394583 Supplier Toronto Research Chemicals Supplier No. TRCH10106025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2033064-44-5
Molecular Formula C24H19N5O7S2
Molecular Weight (g/mol) 553.56
IUPAC Name 2-hydroxy-5-[(E)-[4-[[4-(pyridin-2-ylsulfamoyl)phenyl]sulfamoyl]phenyl]diazenyl]benzoic acid
SMILES OC(=O)c1cc(ccc1O)N=Nc2ccc(cc2)S(=O)(=O)Nc3ccc(cc3)S(=O)(=O)Nc4ccccn4

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C24H19N5O7S2/c30-22-13-8-18(15-21(22)24(31)32)27-26-16-4-9-19(10-5-16)37(33,34)28-17-6-11-20(12-7-17)38(35,36)29-23-3-1-2-14-25-23/h1-15,28,30H,(H,25,29)(H,31,32)
Formula Weight 553.0726
Chemical Name or Material (E)-2-Hydroxy-5-((4-(N-(4-(N-(pyridin-2-yl)sulfamoyl)phenyl)sulfamoyl)phenyl)diazenyl)benzoic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.