missing translation for 'onlineSavingsMsg'
Learn More

(E)-2-Furaldehyde, (2,4-Dinitrophenyl)hydrazone, TRC

Product Code. 30393352
Change view
Click to view available options
Quantity:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30393352 10 mg
30408688 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30393352 Supplier Toronto Research Chemicals Supplier No. TRCF86238510MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16416-22-1
Molecular Formula C11H8N4O5
Molecular Weight (g/mol) 276.2
Synonym (E)-2-Furancarboxaldehyde, (2,4-Dinitrophenyl)hydrazone
IUPAC Name N-[(E)-furan-2-ylmethylideneamino]-2,4-dinitroaniline
SMILES [O-][N+](=O)c1ccc(N\N=C\c2occc2)c(c1)[N+](=O)[O-]

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C11H8N4O5/c16-14(17)8-3-4-10(11(6-8)15(18)19)13-12-7-9-2-1-5-20-9/h1-7,13H/b12-7+
Formula Weight 276.0495
Chemical Name or Material (E)-2-Furaldehyde, (2,4-Dinitrophenyl)hydrazone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.