missing translation for 'onlineSavingsMsg'
Learn More

(E)-1-Pentene-1,2-diboronic acid bis(pinacol) ester, 98%, Thermo Scientific™

Product Code. 11385455
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11385455 5 g 5g
11375455 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11385455 Supplier Thermo Scientific Alfa Aesar Supplier No. L19648.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 307531-75-5
Molecular Formula C17H32B2O4
Molecular Weight (g/mol) 322.06
MDL Number MFCD03093909
InChI Key MXQDNQSRLNMUOP-UHFFFAOYSA-N
Synonym cis-1,2-bis 4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pentene, 1-cis-1,2-bis 4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pentene, 2,2 inverted exclamation marka-1z-1-propyl-1,2-ethenediyl bis 4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4,4,5,5-tetramethyl-2-1z-2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pent-1-en-1-yl-1,3,2-dioxaborolane, 4,4,5,5-tetramethyl-2-z-1-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pent-1-en-2-yl-1,3,2-dioxaborolane
PubChem CID 5900533
SMILES CCCC(=CB1OC(C)(C)C(C)(C)O1)B1OC(C)(C)C(C)(C)O1

Specifications

Boiling Point 301°C to 303°C
Flash Point 76°C (168°F)
Quantity 5 g
IUPAC Name 4,4,5,5-tetramethyl-2-[2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pent-1-en-1-yl]-1,3,2-dioxaborolane
Formula Weight 322.06
Percent Purity 98%
Chemical Name or Material (E)-1-Pentene-1,2-diboronic acid bis(pinacol) ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.