missing translation for 'onlineSavingsMsg'
Learn More

(E)-1-Hexene-1,2-diboronic acid bis(pinacol) ester, 98%, Thermo Scientific™

Product Code. 11315465
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11315465

Brand: Thermo Scientific Alfa Aesar L19650.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 185427-48-9
Molecular Formula C18H34B2O4
Molecular Weight (g/mol) 336.09
MDL Number MFCD03093898,MFCD03093898
InChI Key SACOFNGKSPZAFQ-UHFFFAOYSA-N
Synonym e-1-hexene-1,2-diboronic acid bis pinacol ester, 1e-1,2-bis 4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl hex-1-ene, z-1,2-bis 4,4,5,5-tetramethyl-1,3,2-dioxaborolane-2-yl-1-hexene, 4,4,5,5-tetramethyl-2-1z-2-tetramethyl-1,3,2-dioxaborolan-2-yl hex-1-en-1-yl-1,3,2-dioxaborolane, 4,4,5,5-tetramethyl-2-z-1-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl hex-1-en-2-yl-1,3,2-dioxaborolane, 4,4,5,5-tetramethyl-2-1-tetramethyl-1,3,2-dioxaborolan-2-yl hex-1-en-2-yl-1,3,2-dioxaborolane
PubChem CID 6026893
IUPAC Name 4,4,5,5-tetramethyl-2-[(Z)-1-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)hex-1-en-2-yl]-1,3,2-dioxaborolane
SMILES CCCCC(=CB1OC(C)(C)C(C)(C)O1)B1OC(C)(C)C(C)(C)O1

Specifications

Density 0.954
Boiling Point 102°C to 104°C (0.08 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.456
Quantity 1 g
Beilstein 7768251
Formula Weight 336.09
Percent Purity 98%
Chemical Name or Material (E)-1-Hexene-1,2-diboronic acid bis(pinacol) ester
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.