missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DZIP3 Antibody, PerCP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97549PCP
This item is not returnable.
View return policy
Description
DZIP3 Polyclonal antibody specifically detects DZIP3 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| DZIP3 | |
| Polyclonal | |
| Immunohistochemistry-Paraffin | |
| DAZ interacting protein 3, zinc finger, DAZ-interacting protein 3, E3 ubiquitin-protein ligase DZIP3, EC 6.3.2.-, FLJ13327, FLJ57977, FLJ58022, FLJ58223, hRUL138KIAA0675, human RNA-binding ubiquitin ligase of 138 kDa, RNA-binding ubiquitin ligase of 138 kDa, UURF2, UURF2 ubiquitin ligase, zinc finger DAZ interacting protein 3 | |
| Produced in rabbits immunized with E. coli-derived Human DZIP3 fragment. | |
| 0.1 mL | |
| Zinc Finger | |
| 9666 | |
| Store at 4C in the dark. | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| PerCP | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction