missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ DZIP1L Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA556462

Product Code. 15754371

  • £467.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: LEDTLEEKLE SMGIRKDAKG ISIQTLRHLE SLLRVQREQK ARKFSEFLSL RGKLVKEVTS RAKERQENGA VVSQPDGQPS VKSQQ Highest antigen sequence identity to the following orthologs: Mouse - 67%, Rat - 72%.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

DZIP1L
Polyclonal
Unconjugated
DZIP1L
DAZ interacting protein 1-like; DAZ interacting zinc finger protein 1 like; DAZ interacting zinc finger protein 1-like; DAZ-interacting protein 1-like protein; DZIP1L; DZIP2; Zinc finger protein DZIP1L
Rabbit
Antigen affinity chromatography
RUO
199221
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.2 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q8IYY4
DZIP1L
Recombinant protein corresponding to Human DZIP1L. Recombinant protein control fragment (Product #RP-95520).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.