missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DZIP1L Antibody, Novus Biologicals™
Mouse Polyclonal Antibody
Brand: Novus Biologicals H00199221-B01P
This item is not returnable.
View return policy
Description
DZIP1L Polyclonal antibody specifically detects DZIP1L in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
Specifications
| DZIP1L | |
| Polyclonal | |
| PBS (pH 7.4) | |
| DAZ interacting protein 1-like, DAZ-interacting protein 1-like protein, DZIP2, FLJ32844, zinc finger protein DZIP1L | |
| DZIP1L (NP_775814.1, 1 a.a. - 767 a.a.) full-length human protein. MQSPAATAEGLSGPLFGAYTFPTFKFQPRHDSMDWRRISTLDVDRVARELDVATLQENIAGITFCNLDREVCSRCGQPVDPALLKVLRLAQLIIEYLLHCQDCLSASVAQLEARLQTSLGQQQRGQQELGRQADELKGVREESRRRRKMISTLQQLLMQTGTHSYHTCHLCDKTFMNATFLRGHIQRRHAGVAEGGKQKKQEQPVEEVLEELRAKLKWTQGELEAQREAERQRQLQEAELIHQREIEAKKEFDKWKEQEWTKLYGEIDKLKKLFWDEFKNVAKQNSTLEEKLRALQSHSVMESKLGSLRDEESEEWLRQARELQALREKTEIQKTEWKRKVKELHEEHMAEKKELQEENQRLQASLSQDQKKAAAQSQCQISTLRAQLQEQARIIASQEEMIQSLSLRKVEGIHKVPKAVDTEEDSPEEEMEDSQDEQHKVLAALRRNPTLLKHFRPILEDTLEEKLESMGIRKDAKGISIQTLRHLESLLRVQREQKARKFSEFLSLRGKLVKEVTSRAKERQENGAAVSQPDGQPSVKSQQSALVTREAQPKTRTLQVALPSTPAEPPPPTRQSHGSHGSSLTQVSAPAPHPGLHGPSSTPPSSGPGMSTPPFSSEEDSEGDRVQRVSLQPPKVPSRMVPRPKDDWDWSDTETSEENAQPPGQGSGTLVQSMVKNLEKQLEAPAKKPAGGVSLFFMPNAGPQRAATPGRKPQLSEDESDLEISSLEDLPLDLDQREKPKPLSRSKLPEKFGTGPQSSGQPRVPAW | |
| 0.05 mg | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunocytochemistry/Immunofluorescence | |
| Unconjugated | |
| NP_775814.1 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| 199221 | |
| Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction