missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DZIP1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13779-1-AP-150UL
This item is not returnable.
View return policy
Description
May participate in spermatogenesis via its interaction with DAZ.Specifications
| DZIP1 | |
| Polyclonal | |
| Unconjugated | |
| DZIP1 | |
| DAZ interacting protein 1, DAZ interacting protein 1/2, DZIP, DZIP1, DZIP2, DZIPt1, KIAA0996, RP11 23E3.3, Zinc finger protein DZIP1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 22873, 66573 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q86YF9, Q8BMD2 | |
| Dzip1 | |
| DZIP1 Fusion Protein Ag4772 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction