missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DZIP1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98687-100ul
This item is not returnable.
View return policy
Description
DZIP1 Polyclonal antibody specifically detects DZIP1 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| DZIP1 | |
| Polyclonal | |
| Immunohistochemistry-Paraffin 1:1000-1:4000 | |
| DAZ interacting protein 1, DAZ-interacting protein 1/2, DZIP, DZIP2, DZIPDAZ interacting protein testis1, DZIPt1, KIAA0996zinc finger DAZ interacting protein 1, RP11-23E3.3, zinc finger protein DZIP1, zinc-finger protein DZIPt1 | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human DZIP1. | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 22873 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction