missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DYX1C1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14522-1-AP-20UL
This item is not returnable.
View return policy
Description
This gene encodes a tetratricopeptide repeat domain-containing protein. The encoded protein interacts with estrogen receptors and the heat shock proteins, Hsp70 and Hsp90. A chromosomal translocation involving this gene is associated with a susceptibility to developmental dyslexia. Mutations in this gene are associated with deficits in reading and spelling. Alternate splicing results in multiple transcript variants.Specifications
| DYX1C1 | |
| Polyclonal | |
| Unconjugated | |
| DNAAF4 | |
| DYX1, DYX1C1, DYXC1, EKN1, RD | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 161582, 363096, 67685 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Western Blot | |
| 0.31 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q5VJS5, Q8R368, Q8WXU2 | |
| Dnaaf4 | |
| DYX1C1 Fusion Protein Ag5997 | |
| 20 μL | |
| Primary | |
| Mouse, Rat, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction