Learn More
Invitrogen™ Dystrophin Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5110894
Description
Immunogen sequence: KQNDVHRAFK RELKTKEPVI MSTLETVRIF LTEQPLEGLE KLYQEPRELP PEERAQNVTR LLRKQAEEVN TEWEKLNLHS ADWQRKIDET LERLQELQEA TDELDLKLRQ AEVIKGSWQP VGDLLIDSLQ DHLEKVKALR GEIAPLKENV Highest antigen sequence indentity to the following orthologs: Mouse - 96%, Rat - 98%.
Dystrophin is the 427kDa protein product of the DMB/BMD gene located on the X chromosome at position Xp21. Western blotting and immunohistochemistry are the two established methods for the detection of abnormalities of dystrophin expression in muscle biopsies. Dystrophin abnormalities are thought to occur in 100% of patients with DMD/BMD, although genetic abnormalities may only be detected in up to 65% of cases.
Specifications
| Dystrophin | |
| Polyclonal | |
| Unconjugated | |
| DMD | |
| apodystrophin-3; apodystrophin-I; BMD; CMD3B; dmd; dmd.1; dmd.1.L; DNADMD1; Dp427; Dp71; Duchenne muscular dystrophy (DMD); DXS142; DXS164; DXS206; DXS230; DXS239; DXS268; DXS269; DXS270; DXS272; DXSmh7; DXSmh9; dys; Dystrophin; dystrophin Dp40; dystrophin Dp71 isoform; dystrophin Dp71a; dystrophin Dp71ab; dystrophin Dp71b; dystrophin, gene 1 L homeolog; dystrophin, muscular dystrophy; GS1-19O24.1; mdx; MRX85; Muscular dystrophy Duchenne and Becker types; OTTHUMP00000215592; pke; utrn; utrophin; XELAEV_18012903mg; X-linked muscular dystrophy | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 1756 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P11532 | |
| DMD | |
| Recombinant Protein corresonding to Human Dystrophin. Recombinant protein control fragment (Product #RP-92153). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.