missing translation for 'onlineSavingsMsg'
Learn More

Dystrophin Antibody (DMD/6270), DyLight 550, Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals NBP3-20751R

Product Code. 18557728

  • £600.00 / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Dystrophin Monoclonal antibody specifically detects Dystrophin in Human, Canine samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Dystrophin
Monoclonal
DyLight 550
BMDDXS272, CMD3B, DXS142, DXS164, DXS164, DXS206, DXS230, DXS239, DXS268, DXS269, DXS270, DXS272, DXS206, DXS230, DXS239, DXS268, DXS269, DXS270, dystrophin, dystrophin (muscular dystrophy, Duchenne and Becker types), includes DXS142
Recombinant fragment (around aa1700-2300) of human Dystrophin (exact sequence is proprietary).
0.1 mL
Cell Biology, Cellular Markers
1756
Store at 4°C in the dark.
IgG2a κ
Western Blot, Immunofluorescence, Immunohistochemistry (Paraffin)
DMD/6270
50 mM Sodium Borate
Mouse
Protein A or G purified
RUO
Primary
Human, Canine
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.