missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dystrophin Antibody (DMD/6270), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-20751CL1
This item is not returnable.
View return policy
Description
Dystrophin Monoclonal antibody specifically detects Dystrophin in Canine,Human samples. It is validated for Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/Immunofluorescence
Specifications
| Dystrophin | |
| Monoclonal | |
| CoraFluor 1 | |
| BMDDXS272, CMD3B, DXS142, DXS164, DXS164, DXS206, DXS230, DXS239, DXS268, DXS269, DXS270, DXS272, DXS206, DXS230, DXS239, DXS268, DXS269, DXS270, dystrophin, dystrophin (muscular dystrophy, Duchenne and Becker types), includes DXS142 | |
| Recombinant fragment (around aa1700-2300) of human Dystrophin (exact sequence is proprietary). | |
| 0.1 mL | |
| Cell Biology, Cellular Markers | |
| 1756 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG2a κ |
| Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence | |
| DMD/6270 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Canine, Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction