missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dystrophin Antibody (DMD/3241), Alexa Fluor™ 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-79910AF488
This item is not returnable.
View return policy
Description
Dystrophin Monoclonal specifically detects Dystrophin in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Dystrophin | |
| Monoclonal | |
| Alexa Fluor 488 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| DMD | |
| A recombinant fragment (around aa 114-263) of human DMD protein (exact sequence is proprietary) (Uniprot: P11532) | |
| 0.1 mL | |
| Cell Biology, Cellular Markers | |
| Human | |
| IgG1 κ |
| ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| DMD/3241 | |
| ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| BMDDXS272, CMD3B, DXS142, DXS164, DXS164, DXS206, DXS230, DXS239, DXS268, DXS269, DXS270, DXS272, DXS206, DXS230, DXS239, DXS268, DXS269, DXS270, dystrophin, dystrophin (muscular dystrophy, Duchenne and Becker types), includes DXS142 | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction