missing translation for 'onlineSavingsMsg'
Learn More
Learn More
dystrophia myotonica containing WD repeat motif Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£180.00 - £248.00
Specifications
| Antigen | dystrophia myotonica containing WD repeat motif |
|---|---|
| Dilution | Immunoprecipitation 0.5-2 μL/mg of lysate |
| Applications | Immunoprecipitation |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18644602
|
Novus Biologicals
NBP2-97522-100ul |
100 μg |
£248.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18615714
|
Novus Biologicals
NBP2-97522-50ul |
£180.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
Description
dystrophia myotonica containing WD repeat motif Polyclonal antibody specifically detects dystrophia myotonica containing WD repeat motif in Human samples. It is validated for ImmunoprecipitationSpecifications
| dystrophia myotonica containing WD repeat motif | |
| Immunoprecipitation | |
| Unconjugated | |
| Rabbit | |
| Cell Cycle and Replication | |
| PBS (pH 7.0) | |
| 1762 | |
| IgG | |
| Antigen and protein A Affinity-purified |
| Immunoprecipitation 0.5-2 μL/mg of lysate | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| D19S593E, DM9, DMRN9, DMR-N9, dystrophia myotonica WD repeat-containing protein, dystrophia myotonica, WD repeat containing, dystrophia myotonica-containing WD repeat motif, Dystrophia myotonica-containing WD repeat motif protein, gene59, Protein 59, Protein DMR-N9 | |
| Produced in rabbits immunized with E. coli-derived Human dystrophia myotonica containing WD repeat motif fragment. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title