missing translation for 'onlineSavingsMsg'
Learn More

Dystrobrevin alpha Antibody (OTI1B2), DyLight 550, Novus Biologicals™

Product Code. 18171342 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18171342 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18171342 Supplier Novus Biologicals Supplier No. NBP270591R

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Dystrobrevin alpha Monoclonal specifically detects Dystrobrevin alpha in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Dystrobrevin alpha
Applications Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone OTI1B2
Conjugate DyLight 550
Dilution Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias Alpha-dystrobrevin, D18S892EDTN, DRP3FLJ96209, DTN-1, DTN-2, DTN-3, DTN-A, dystrobrevin alpha, dystrobrevin, alpha, Dystrophin-related protein 3, LVNC1
Gene Symbols DTNA
Host Species Mouse
Immunogen Full length recombinant protein of human Dystrobrevin alpha (NP_116761) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 1837.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.