missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dystonin Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£145.00 - £417.00
Specifications
| Antigen | Dystonin |
|---|---|
| Dilution | Western Blot 1:500 - 1:1000, ELISA |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30230960
|
Novus Biologicals
NBP3-35915-100ul |
100 μL |
£417.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30229342
|
Novus Biologicals
NBP3-35915-20ul |
20 μL |
£145.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Dystonin Polyclonal antibody specifically detects Dystonin in Human,Mouse,Rat samples. It is validated for ELISA,Western BlotSpecifications
| Dystonin | |
| ELISA, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cancer | |
| PBS (pH 7.3), 50% glycerol | |
| 667 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500 - 1:1000, ELISA | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| 230 kDa bullous pemphigoid antigen, BP240, BPAFLJ30627, BPAG1FLJ32235, Bullous pemphigoid antigen, Bullous pemphigoid antigen 1, bullous pemphigoid antigen 1, 230/240kDa, CATX-15, D6S1101, DKFZp564B2416, DMHCATX15, DT, Dystonia musculorum protein, dystonin, FLJ13425, FLJ21489, FLJ46791, Hemidesmosomal plaque protein, KIAA0465, KIAA0728, KIAA1470, MACF2, trabeculin-beta | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-270 of human Dystonin (NP_001714.1).,, Sequence:, MHSSSYSYRSSDSVFSNTTSTRTSLDSNENLLLVHCGPTLINSCISFGSESFDGHRLEMLQQIANRVQRDSVICEDKLILAGNALQSDSKRLESGVQFQNEAEIAGYILECENLLRQHVIDVQILIDGKYYQADQLVQRVAKLRDEIMALRNECSSVYSKGRILTTEQTKLMISGITQSLNSGFAQTLHPSLTSGLTQSLTPSLTSSSMTSGLSSGMTSRLTPSVTPAYTPGFPSGLVPNFSSGVEPNSLQTLKLMQIRKPLLKSSLLDQ | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title