missing translation for 'onlineSavingsMsg'
Learn More

Dysprosium rod, 12.7mm (0.5in) dia, 99.9% (metals basis excluding Ta)

Product Code. 11353808
Change view
Click to view available options
Quantity:
10 mm
25 mm
50 mm
100 mm
Unit Size:
100mm
10mm
25mm
50mm
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11353808 25 mm 25mm
11383808 10 mm 10mm
11373808 50 mm 50mm
11363808 100 mm 100mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11353808 Supplier Thermo Scientific Alfa Aesar Supplier No. 040286.DH
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 7429-91-6 | Dy | 162.50 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7429-91-6
Molecular Formula Dy
Molecular Weight (g/mol) 162.50
MDL Number MFCD00010982
InChI Key KBQHZAAAGSGFKK-UHFFFAOYSA-N
Synonym ingot reo, disprosio, atom, ion, ingot, pieces, powder, foil, 3n, chips, 3n, ingot, 3n
PubChem CID 23912
ChEBI CHEBI:33377
IUPAC Name dysprosium
SMILES [Dy]

Specifications

Quantity 25 mm
Odor Odorless
Packaging Mylar under Argon
Weight ≈1.1g/mm
Form Cast Rod
Percent Purity 99.9%
Assay (metals basis excluding Ta)
Chemical Name or Material Dysprosium rod, 12.7mm (0.5 in.) dia.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.