missing translation for 'onlineSavingsMsg'
Learn More

Dysprosium pieces, 99.9% (REO), Thermo Scientific Chemicals

Catalog Number 11350659
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy
7429-91-6
Dy
162.50
MFCD00010982
KBQHZAAAGSGFKK-UHFFFAOYSA-N
ingot reo, disprosio, atom, ion, ingot, pieces, powder, foil, 3n, chips, 3n, ingot, 3n
23912
CHEBI:33377
dysprosium
[Dy]
This item is not returnable. View return policy
7429-91-6
Dy
162.50
MFCD00010982
KBQHZAAAGSGFKK-UHFFFAOYSA-N
ingot reo, disprosio, atom, ion, ingot, pieces, powder, foil, 3n, chips, 3n, ingot, 3n
23912
CHEBI:33377
dysprosium
[Dy]

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under argon
TRUSTED_SUSTAINABILITY
Color Silver to Gray
UN Number UN3178
Quantity 50 g
Percent Purity 99.9%
Odor Odorless
Form Pieces
Assay (REO)
Chemical Name or Material Dysprosium pieces

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'