missing translation for 'onlineSavingsMsg'
Learn More

Dysprosium(III) oxalate decahydrate, REacton™, 99.9% (REO), Thermo Scientific™

Product Code. 11382538
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11382538 100 g 100g
11392538 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11382538 Supplier Thermo Scientific Alfa Aesar Supplier No. 014593.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24670-07-3
Molecular Formula C6H26Dy2O22
Molecular Weight (g/mol) 775.252
MDL Number MFCD00150482
InChI Key WJYBVSNZIQHDSA-UHFFFAOYSA-N
Synonym scandium oxalate decahydrate
PubChem CID 131876564
IUPAC Name dysprosium;oxalic acid;decahydrate
SMILES C(=O)(C(=O)O)O.C(=O)(C(=O)O)O.C(=O)(C(=O)O)O.O.O.O.O.O.O.O.O.O.O.[Dy].[Dy]

Specifications

Melting Point 40°C (in -H2O)
Color White
Physical Form Powder
Quantity 100 g
Linear Formula Dy2(C2O4)3·10H2O
UN Number UN3288
Formula Weight 769.21 (589.06 Anhydrous)
Percent Purity 99.9%
Grade Reacton
Sensitivity Hygroscopic
Assay (REO)
Chemical Name or Material Dysprosium(III) oxalate decahydrate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.