missing translation for 'onlineSavingsMsg'
Learn More

Dysprosium(III) isopropoxide, 99.9% (REO), 5% w/v in toluene/isopropanol

Product Code. 11398018
Change view
Click to view available options
Quantity:
5 mL
25 mL
100 mL
Unit Size:
100mL
25mL
5mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11398018 5 mL 5mL
11318028 25 mL 25mL
11308028 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11398018 Supplier Thermo Scientific Alfa Aesar Supplier No. 036585.AA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 6742-68-3 | C9H24DyO3 | 342.788 g/mol

Dysprosium(III) isopropoxide is used as primary and secondary intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6742-68-3
Molecular Formula C9H24DyO3
Molecular Weight (g/mol) 342.788
MDL Number MFCD00049784
InChI Key IWDABVYEISORRX-UHFFFAOYSA-N
Synonym dysprosium iii i-propoxide-dy reo
PubChem CID 131675877
IUPAC Name dysprosium;propan-2-ol
SMILES CC(C)O.CC(C)O.CC(C)O.[Dy]

Specifications

Color Brown
Linear Formula Dy[OCH(CH3)2]3
Quantity 5 mL
UN Number UN1993
Sensitivity Moisture sensitive
Solubility Information Insoluble in water.
Formula Weight 339.76
Percent Purity 99.9%
Concentration or Composition (by Analyte or Components) 5% w/v in toluene/isopropanol
Physical Form Liquid
Assay (REO)
Chemical Name or Material Dysprosium(III) isopropoxide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.