missing translation for 'onlineSavingsMsg'
Learn More

Dysprosium(III) iodide, ultra dry, 99.99% (REO)

Product Code. 11353877
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11353877 1 g 1g
11343877 5 g 5g
11363877 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11353877 Supplier Thermo Scientific Alfa Aesar Supplier No. 035693.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 15474-63-2 | DyI3 | 543.213 g/mol

It is used in high-intensity metal-halide lamps.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15474-63-2
Molecular Formula DyI3
Molecular Weight (g/mol) 543.213
MDL Number MFCD00016072
InChI Key RZQFCZYXPRKMTP-UHFFFAOYSA-K
Synonym dysprosium iii iodide, dysprosium triiodide, dysprosium iodide, acmc-20al3a, dysprosium iii ; iodide, dysprosium 3+ triiodide, dysprosium iii iodide, anhydrous
PubChem CID 84931
IUPAC Name dysprosium(3+);triiodide
SMILES [I-].[I-].[I-].[Dy+3]

Specifications

Melting Point 955°C
Color Green to Yellow
Physical Form Flakes
Quantity 1 g
Solubility Information Soluble in water.
Formula Weight 543.21
Percent Purity 99.99%
Packaging Ampule
Sensitivity Hygroscopic
Assay (REO)
Chemical Name or Material Dysprosium(III) iodide, Ultra dry
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.