missing translation for 'onlineSavingsMsg'
Learn More

Dysprosium(III) fluoride, anhydrous, 99.9% (REO), Thermo Scientific™

Product Code. 11384108
Change view
Click to view available options
Quantity:
50 g
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11384108 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11384108 Supplier Thermo Scientific Alfa Aesar Supplier No. 041409.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

For vacuum deposition

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13569-80-7
MDL Number MFCD00016071

Specifications

Boiling Point 2200°C
Melting Point 1360°C
Color White
Physical Form Pieces
Quantity 50 g
Solubility Information Soluble in strong mineral acids. Insoluble in water.
Formula Weight 219.5
Percent Purity 99.9%
Odor Odorless
Sensitivity Hygroscopic
Density 7.5 g/mL
Assay (REO)
Chemical Name or Material Dysprosium(III) fluoride, Anhydrous
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.