missing translation for 'onlineSavingsMsg'
Learn More

Dysprosium(III) chloride, ultra dry, 99.98% (REO)

Product Code. 11323877
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11323877

Brand: Thermo Scientific Alfa Aesar 035692.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 10025-74-8 | Cl3Dy | 268.85 g/mol

It is used as a precursor for the preparation of other dysprosium salts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10025-74-8
Molecular Formula Cl3Dy
Molecular Weight (g/mol) 268.85
MDL Number MFCD00010984
InChI Key BOXVSFHSLKQLNZ-UHFFFAOYSA-K
IUPAC Name dysprosium(3+) trichloride
SMILES [Cl-].[Cl-].[Cl-].[Dy+3]

Specifications

Melting Point 718°C
Physical Form Powder
Quantity 25 g
Merck Index 14,3482
Solubility Information Soluble in water.
Formula Weight 268.86
Percent Purity 99.98%
Odor Odorless
Packaging Ampule
Sensitivity Hygroscopic
Density 3.67 g/mL
Assay (REO)
Chemical Name or Material Dysprosium(III) chloride, Ultra dry
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.