missing translation for 'onlineSavingsMsg'
Learn More

Dysprosium(III) chloride hydrate, 99.99% (REO)

Product Code. 11361069
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11361069 100 g 100g
11341069 5 g 5g
11351069 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11361069 Supplier Thermo Scientific Alfa Aesar Supplier No. 011317.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

MDL Number MFCD00010984

Specifications

Physical Form Wet Crystalline Aggregates
Quantity 100 g
Formula Weight 268.86 (Anhydrous)
Percent Purity 99.99%
Sensitivity Hygroscopic
Assay (REO)
Chemical Name or Material Dysprosium(III) chloride hydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.