missing translation for 'onlineSavingsMsg'
Learn More

Dysprosium(III) carbonate tetrahydrate, 99.9% (REO)

Product Code. 11357108
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
This item is not returnable. View return policy

Product Code. 11357108

Brand: Thermo Scientific Alfa Aesar 015286.30

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 38245-35-1 | C3H8Dy2O13 | 577.084 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 38245-35-1
Molecular Formula C3H8Dy2O13
Molecular Weight (g/mol) 577.084
MDL Number MFCD00150229
InChI Key BQRZSEZQNZPPPU-UHFFFAOYSA-H
Synonym dysprosium carbonate tetrahydrate, acmc-20al3d, dysprosium carbonate, dysprosium iii carbonate tetrahydrate, dysprosium carbonate-water 2/3/4, didysprosium 3+ ion tetrahydrate trico3
PubChem CID 25021600
IUPAC Name dysprosium(3+);tricarbonate;tetrahydrate
SMILES C(=O)([O-])[O-].C(=O)([O-])[O-].C(=O)([O-])[O-].O.O.O.O.[Dy+3].[Dy+3]

Specifications

Physical Form Powder
Quantity 250 g
Formula Weight 577.06 (505.03 Anhydrous)
Percent Purity 99.9%
Sensitivity Hygroscopic
Assay (REO)
Chemical Name or Material Dysprosium(III) carbonate tetrahydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.