missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dysferlin Antibody (CL10888) [mFluor Violet 450 SE], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-07994MFV450
This item is not returnable.
View return policy
Description
Dysferlin Monoclonal antibody specifically detects Dysferlin in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
Specifications
| Dysferlin | |
| Monoclonal | |
| mFluor Violet 450 SE | |
| dysferlin, dysferlin, limb girdle muscular dystrophy 2B (autosomal recessive), dystrophy-associated fer-1-like 1, FER1L1Dystrophy-associated fer-1-like protein, Fer-1-like protein 1, FLJ00175, FLJ90168, LGMD2B | |
| This antibody was developed using a recombinant protein derived from O75923, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Cell Biology, Cellular Markers | |
| 8291 | |
| Store at 4⪚C in the dark. | |
| IgG1 |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
| CL10888 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction