missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dysbindin Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35661-100ul
This item is not returnable.
View return policy
Description
Dysbindin Polyclonal antibody specifically detects Dysbindin in Human,Mouse samples. It is validated for ELISA,Western Blot
Specifications
| Dysbindin | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA | |
| DKFZp564K192, dysbindin, Dysbindin-1, dystrobrevin binding protein 1, Dystrobrevin-binding protein 1, FLJ30031, Hermansky-Pudlak syndrome 7 protein, HPS7 protein, HPS7DBND, MGC20210, My031, SDY | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-351 of human Dysbindin (NP_115498.2).,, Sequence:, MLETLRERLLSVQQDFTSGLKTLSDKSREAKVKSKPRTVPFLPKYSAGLELLSRYEDTWAALHRRAKDCASAGELVDSEVVMLSAHWEKKKTSLVELQEQLQQLPALIADLESMTANLTHLEASFEEVENNLLHLEDLCGQCELERCKHMQSQQLENYKKNKRKELETFKAELDAEHAQKVLEMEHTQQMKLKERQKFFEEAFQQDMEQYLSTGYLQIAERREPIGSMSSMEVNVDMLEQMDLMDISDQEALDVFLNSGGEENTVLSPALGPESSTCQNEITLQVPNPSELRAKPPSSSSTCTDSATRDISEGGESPVVQSDEEEVQVDTALATSHTDREATPDGGEDSDS | |
| 100 μL | |
| Neuroscience | |
| 84062 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction