missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Dysadherin Monoclonal Antibody (M029)

Description
Clone M029 mouse monoclonal antibody detects a 35-55 kDa* band on SDS-PAGE immunoblots of human A431, A549, MeWo, MDA-MB-231 cell, as well as human breast, lung, and skin tissues. The antibody works for western blot, immunoprecipitation, ELISA, and immunocytochemistry, as well as detects the FXYD5 protein on live cells.
This gene encodes a member of a family of small membrane proteins that share a 35-amino acid signature sequence domain, beginning with the sequence PFXYD and containing 7 in variant and 6 highly conserved amino acids. The approved human gene nomenclature for the family is FXYD-domain containing ion transport regulator. Mouse FXYD5 has been termed RIC (Related to Ion Channel). FXYD2, also known as the gamma subunit of the Na,K-ATPase, regulates the properties of that enzyme. FXYD1 (phospholemman), FXYD2 (gamma), FXYD3 (MAT-8), FXYD4 (CHIF), and FXYD5 (RIC) have been shown to induce channel activity in experimental expression systems. Transmembrane topology has been established for two family members (FXYD1 and FXYD2), with the N-terminus extracellular and the C-terminus on the cytoplasmic side of the membrane. This gene product, FXYD5, is a glycoprotein that functions in the up-regulation of chemokine production, and it is involved in the reduction of cell adhesion via its ability to down-regulate E-cadherin. It also promotes metastasis, and has been linked to a variety of cancers. Alternative splicing results in multiple transcript variants.
Specifications
Specifications
| Antigen | Dysadherin |
| Applications | Immunoprecipitation, Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | M029 |
| Concentration | 0.25 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1mg/mL BSA, 50% glycerol and 0.05% sodium azide |
| Gene | FXYD5 |
| Gene Accession No. | Q96DB9 |
| Gene Alias | DYSAD; Dysadherin; EF-8; FXYD domain containing ion transport regulator 5; FXYD domain-containing ion transport regulator 5; FXYD5; HSPC113; Ion channel homolog RIC; IWU1; IWU-1; KCT1; keratinocytes associated transmembrane protein 1; OIT2; oncoprotein induced transcript 2; oncoprotein-induced protein 2; PRO6241; RIC; UNQ2561/PRO6241 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?