missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DYRK4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12890-2-AP-150UL
This item is not returnable.
View return policy
Description
Dual-specificity kinases, such as DYRK4, play key roles in cell proliferation, survival, and development.Specifications
| DYRK4 | |
| Polyclonal | |
| Unconjugated | |
| DYRK4 | |
| DYRK4 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 101320, 312721, 8798 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.23 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q8BI55, Q9NR20 | |
| Dyrk4 | |
| DYRK4 Fusion Protein Ag3515 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction