missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DYRK3 Monoclonal Antibody (563518)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA561549
This item is not returnable.
View return policy
Description
Reconstitute at 0.5 mg/mL in sterile PBS.
DYRK3 belongs to the DYRK family of dual-specificity protein kinases that catalyze phosphorylation of serine/threonine and tyrosine residues. The members of this family share structural similarity, but differ in their substrate specificity, suggesting their involvement in different cellular functions. DYRK3 is a negative regulator of EPO-dependent erythropoiesis, may place an upper limit on red cell production during stress erythropoiesis. Inhibits cell death due to cytokine withdrawal in hematopoietic progenitor cells. May act by regulating CREB/CRE signaling.
Specifications
| DYRK3 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with trehalose and no preservative | |
| O43781, Q4V8A3, Q922Y0 | |
| DYRK3 | |
| E. coli-derived recombinant human DYRK3, Asn52-Glu167. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG1 |
| Western Blot | |
| 563518 | |
| Unconjugated | |
| DYRK3 | |
| BC006704; dual specificity tyrosine phosphorylation regulated kinase 3; dual specificity tyrosine-(Y)-phosphorylation regulated kinase 3; dual specificity tyrosine-phosphorylation-regulated kinase 3; dual-specificity tyrosine-(Y)-phosphorylation regulated kinase 3; dual-specificity tyrosine-(Y)-phosphorylation regulated kinase 5; dyrk3; dyrk3 {ECO:0000312; DYRK5; EMBL:AAH97474.1}; hYAK3-2; KSR; protein kinase Dyrk3; RED; REDK; regulatory erythroid kinase; RP11-343H5.2; RSU2 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 226419, 304775, 8444 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction