missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DYRK2 Polyclonal Antibody

Sheep Polyclonal Antibody
Brand: Invitrogen™ PA547734
This item is not returnable.
View return policy
Description
This antibody detects endogenous human, mouse and rat DYRK2 in Western blots. Reconstitute at 0.2 mg/mL in sterile PBS.
DYRK2 (also known as dual-specificity tyrosine-(Y)-phosphorylation regulated kinase 2) belongs to a family of protein kinases whose members are presumed to be involved in cellular growth and/or development and is defined by structural similarity of their kinase domains and their capability to autophosphorylate on tyrosine residues. DYRK2 expressed in E. coli demonstrated tyrosine autophosphorylation and catalyzed phosphorylation of histones H3 and H2B in vitro. DYRK2 also regulates p53 to induce apoptosis in response to DNA damage.
Specifications
| DYRK2 | |
| Polyclonal | |
| Unconjugated | |
| DYRK2 | |
| 1810038L18Rik; dual specificity tyrosine phosphorylation regulated kinase 2; dual specificity tyrosine-(Y)-phosphorylation regulated kinase 2; dual specificity tyrosine-phosphorylation-regulated kinase 2; dual-specificity tyrosine-(Y)-phosphorylation regulated kinase 2; dyrk2 | |
| Sheep | |
| Antigen Affinity Chromatography | |
| RUO | |
| 314862, 69181, 8445 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Western Blot | |
| 0.2 mg/mL | |
| PBS with 5% trehalose and No Preservative | |
| Q5U4C9, Q92630 | |
| DYRK2 | |
| E. coli-derived recombinant human DYRK2 Met74-Glu180. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction