missing translation for 'onlineSavingsMsg'
Learn More

DYRK2 Antibody, Alexa Fluor™ 405, Novus Biologicals™

Product Code. 18423406 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18423406 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18423406 Supplier Novus Biologicals Supplier No. NBP176560AF405

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

DYRK2 Polyclonal antibody specifically detects DYRK2 in Human samples. It is validated for Western Blot, ELISA, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen DYRK2
Applications Western Blot, ELISA, Immunofluorescence
Classification Polyclonal
Conjugate Alexa Fluor 405
Formulation 50mM Sodium Borate
Gene Alias dual specificity tyrosine-phosphorylation-regulated kinase 2, dual-specificity tyrosine-(Y)-phosphorylation regulated kinase 2, EC 2.7.12, EC 2.7.12.1, FLJ21217, FLJ21365
Host Species Rabbit
Immunogen Antibody was raised against an 18 amino acid synthetic peptide near the carboxy terminus of human DYRK2. The immunogen is located within amino acids 520 - 570 of DYRK2. Amino Acid Squence: LRRRLPKPPTGEKTSCKR
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 8445
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.